missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human LILRA3/CD85e Fc Chimera Protein

Product Code. 18788619
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18788619

Brand: R&D Systems™ 9517T4100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human LILRA3/CD85e Fc Chimera Protein is derived from HEK293. The Recombinant Human LILRA3/CD85e Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N6C8-1
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 11026
Molecular Weight (g/mol) 72kDa
Quantity 100 μg
Source Human embryonic kidney cell,HEK293-derived human LILRA3/CD85e protein, (N-terminus) Human LILRA3/CD85e (Gly24-Glu439) Accession &Num; Q8N6C8-1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias CD85 antigen-like family member E, CD85e, CD85e antigen, HM31, HM43, ILT6, ILT-6, ILT6CD85e, Immunoglobulin-like transcript 6, leucocyte immunoglobulin-like receptor, Leukocyte immunoglobulin-like receptor 4, leukocyte immunoglobulin-like receptor A3, leukocyte immunoglobulin-like receptor subfamily A member 3, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member3, LIR4, LIR4CD85E, LIR-4HM43e3, Monocyte inhibitory receptor HM43/HM31
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.