missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human LIF Protein

Product Code. 18319707
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
100µg
25µg
500µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Formulation unitSize
18319707 25 μg Lyophilized from a 0.2 μm filtered solution in PBS. 25µg
18147750 25 μg Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. 25µg
18187980 100 μg Lyophilized from a 0.2 μm filtered solution in PBS. 100µg
18147550 100 μg Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. 100µg
18736118 500 μg Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. 500µg
18708317 500 μg Lyophilized from a 0.2 μm filtered solution in PBS. 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 18319707 Supplier R&D Systems™ Supplier No. 7734LF025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 0.02-0.12 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P15018
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3976
Molecular Weight (g/mol) M.W. (Observed): 19 kDa, reducing conditions; M.W. (theoretical): 19.6 kDa
Name LIF
Quantity 25 μg
Source E. coli-derived human LIF protein Pro24-Phe202
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Gene Symbol LIF
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein LIF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.