missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Leukotriene A4 Hydrolase/LTA4H Protein

Product Code. 18365317
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365317 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18365317 Supplier R&D Systems™ Supplier No. 4008ZN010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave the fluorogenic peptide substrate, Arg-7-amido-4-methylcoumarin (R-AMC). The specific activity is >70 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09960
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, and Glycerol.
Gene ID (Entrez) 4048
Molecular Weight (g/mol) M.W. (Observed): 71 kDa, reducing conditions; M.W. (theoretical): 70 kDa
Name Leukotriene A4 Hydrolase/LTA4H
Quantity 10 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Leukotriene A4 Hydrolase/LTA4H protein Pro2-Asp611, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol LTA4H
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Leukotriene A4 Hydrolase/LTA4H
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.