missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human LEDGF p52 Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Human LEDGF p52 Protein is derived from E. coli. The Recombinant Human LEDGF p52 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 11168 |
| Molecular Weight (g/mol) | 37.7 kDa |
| Name | LEDGF p52 |
| Quantity | 50 μg |
| Source | E. coli-derived human LEDGF protein Met1-Gln333 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction