missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LCN1 Protein
A cDNA sequence encoding the LCN1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12545-20ug
Additional Details : Weight : 0.01000kg
Specifications
3933 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMHHLLA SDEEIQDVSG TWYLKAMTVD REFPEMNLES VTPMTLTTLE GGNLEAKVTM LISGRCQEVK AVLEKTDEPG KYTADGGKHV AYIIRSHVKD HYIFYCEGEL HGKPVRGVKL VGRDPKNNLE ALEDFEKAAG ARGLSTESIL IPRQSETCSP GSD | |
Greater than 90.0% as determined by SDS-PAGE. |
LCN1 Protein | |
Research Use Only | |
LCN1 | |
Human | |
His | |
LCN1 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |