missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Latent TGF-beta 1 Protein

Product Code. p-4720201
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Supplied as a 0.2μm filtered solution in PBS and Glycerol with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in PBS and Glycerol.
Unit Size:
25µg
5µg
This item is not returnable. View return policy

Product Code. 18127290

Brand: R&D Systems™ 299LT005

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Latent TGF-beta 1 Protein is derived from CHO. The Recombinant Human Latent TGF-beta 1 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 0.2μm filtered solution in PBS and Glycerol with BSA as a carrier protein.
Gene ID (Entrez) 7040
Molecular Weight (g/mol) Observed Molecular Wt: 40-43kDa & 12kDa, reducing conditions 80-90kDa, non-reducing conditions, Predicted Molecular Wt.: 41.3kDa (monomer)
Name Latent TGF-beta 1
Quantity 5 μg
Source Chinese Hamster Ovary cell line,CHO-derived human TGF-beta 1 protein Leu30-Ser390
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, ≤ -20° C under sterile conditions after opening.
Termination Leu30 & Ala279
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.