missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human LAR Protein

Product Code. 18735698
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18735698

Brand: R&D Systems™ 9377PF050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Human LRFN5 Fc Chimera (Catalog # 9385-SA ) is used at 1 μg/mL, the concentration of Recombinant Human LAR that produces 50% optimal binding response is 1.5-9 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P10586-2
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA and Trehalose.
Gene ID (Entrez) 5792
Molecular Weight (g/mol) M.W. (Observed): 114-146 kDa and 14 kDa, reducing conditions; M.W. (theoretical): 136 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human LAR protein Ala27-Glu1251 with a C-terminal 6-His tag, and its proteolyzed forms Ala27-Arg1169 and Gln1170-Glu1251 with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias LCA-homolog, protein tyrosine phosphatase, receptor type, F, PTPRF, receptor type, F polypeptide, receptor-linked protein-tyrosine phosphatase LAR, receptor-type tyrosine-protein phosphatase F
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein LAR/PTPRF
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.