missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human L2HGDH, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) L2HGDH, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8068-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022481

  • £1735.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_079160.1
L2HGDH, mitochondrial Protein
Research Use Only
L2HGDH
Recombinant Protein
E. coli
VIVGGGIVGL ASARALILRH PSLSIGVLEK EKDLAVHQTG HNSGVIHSGI YYKPESLKAK LCVQGAALLY EYCQQKGISY KQCGKLIVAV EQEEIPRLQA LYEKGLQNGV PGLRLIQQED IKKKEPYCRG LMAIDCPHTG IVDYRQVALS FAQDFQEAGG SVLTNFEVKG IEMAKESPSR SIDGMQYPIV IKNTKGEEIR CQYVVTCAGL YSDRISELSG CTPDPRIVPF RGDYLLLKPE KCYLVKGNIY PVPDSRFPFL GVHFTPRMDG SIWLGPNAVL AFKREGYRPF DFSATDVMDI IINSGLIKLA SQNFSYGVTE MYKACFLGAT VKYLQKFIPE ITISDILRGP AGVRAQALDR DGNLVEDFVF DAGVGDIGNR ILHVRNAPSP AATSSIAISG MIADEVQQRF EL
Greater than 90% as determined by SDS-PAGE.
79944
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers