missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLRB1 Protein
Click to view available options
Quantity:
1 μg
1 mg
20 μg
5 μg
50 μg
Unit Size:
1mg
1µg
20µg
50µg
5µg
Specifications
Specifications
| Name | KLRB1 Protein |
| Quantity | 1 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMQKSSIE KCSVDIQQSR NKTTERPGLL NCPIYWQQLR EKCLLFSHTV NPWNNSLADC STKESSLLLI RDKDELIHTQ NLIRDKAILF WIGLNFSLSE KNWKWINGSF LNSNDLEIRG DAKENSCISI SQTSVYSEYC STEIRWICQK ELTPVRNKVY PDS |
| Buffer | The KLRB1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 4M urea and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction