missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Klotho (aa 34-981) Protein

Product Code. 18118530
Change view
Click to view available options
25ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18118530 25ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18118530 Supplier R&D Systems™ Supplier No. 5334KL025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with human FGF RIIIc. The ED50 for this effect is 0.025-0.1 μg/mL in the presence of Recombinant Human FGF-23 (Catalog # 2604-FG ) and Heparin.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UEF7
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in MES, Glycerol and EDTA.
Gene ID (Entrez) 9365
Molecular Weight (g/mol) M.W. (Observed): 120-130 kDa, reducing conditions; M.W. (theoretical): 109.8 kDa
Name Klotho
Quantity 25 μg
Source Mouse myeloma cell line, NS0-derived human Klotho protein Glu34-Ser981, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol KL
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Protein Klotho
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.