missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KLK3 Protein

A cDNA sequence encoding the KLK3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12506-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960639

  • £1210.00 / 100µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

354
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Human
IVGGWECEKH SQPWQVLVAS RGRAVCGGVL VHPQWVLTAA HCIRNKSVIL LGRHSLFHPE DTGQVFQVSH SFPHPLYDMS LLKNRFLRPG DDSSHDLMLL RLSEPAELTD AVKVMDLPTQ EPALGTTCYA SGWGSIEPEE FLTPKKLQCV DLHVISNDVC AQVHPQKVTK FMLCAGRWTG GKSTCSGDSG GPLVCNGVLQ GITSWGSEPC ALPERPSLYT KVVHYRKWIK DTIVANP
Greater than 90% as determined by SDS-PAGE.
KLK3 Protein
Research Use Only
KLK3
Human
Untagged
KLK3 protein filtered (0.4microm) and lyophilized in 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl pH 7.4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers