missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLK13 Protein
Click to view available options
Quantity:
1 μg
5 μg
50 μg
Unit Size:
1µg
50µg
5µg
Specifications
Specifications
| Name | KLK13 Protein |
| Quantity | 1 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSGGVSQES SKVLNTNGTS GFLPGGYTCF PHSQPWQAAL LVQGRLLCGG VLVHPKWVLT AAHCLKEGLK VYLGKHALGR VEAGEQVREV VHSIPHPEYR RSPTHLNHDH DIMLLELQSP VQLTGYIQTL PLSHNNRLTP GTTCRVSGWG TTTSPQVNYP KTLQCANIQL RSDEECRQVY PGKITDNMLC AGTKEGGKDS CEGDSGGPLV CNRTLYGIVS WGDFPCGQPD RPGVYTRVSR YVLWIRETIR KYETQQQKWL KGPQ |
| Buffer | KLK13 protein solution (0.25 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction