missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KLK13 Protein

A cDNA sequence encoding the KLK13 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10737-1ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935018

  • £99.15 / 1µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

KLK13 Protein
Research Use Only
Specific activity is > 8,000 pmol/min/ug. One unit will hydrolyze 1.0 pmole of BAEE to Na-Benzoyl-L-arginine per minute at pH8.0 at 25C.
Human
His
KLK13 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
1 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Baculovirus
GGVSQESSKV LNTNGTSGFL PGGYTCFPHS QPWQAALLVQ GRLLCGGVLV HPKWVLTAAH CLKEGLKVYL GKHALGRVEA GEQVREVVHS IPHPEYRRSP THLNHDHDIM LLELQSPVQL TGYIQTLPLS HNNRLTPGTT CRVSGWGTTT SPQVNYPKTL QCANIQLRSD EECRQVYPGK ITDNMLCAGT KEGGKDSCEG DSGGPLVCNR TLYGIVSWGD FPCGQPDRPG VYTRVSRYVL WIRETIRKYE TQQQKWLKGP QHHHHHH
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers