missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLF7 Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | KLF7 Protein |
| Quantity | 20 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMDVLASY SIFQELQLVH DTGYFSALPS LEETWQQTCL ELERYLQTEP RRISETFGED LDCFLHASPP PCIEESFRRL DPLLLPVEAA ICEKSSAVDI LLSRDKLLSE TCLSLQPASS SLDSYTAVNQ AQLNAVTSLT PPSSPELSRH LVKTSQTLSA VDGTVTLKLV AKKAALSSVK VGGVATAAAA VTAAGAVKSG QSDSDQGGLG AEACPENKKR VHRCQFNGCR KVYTKSSHLK AHQRTHTGEK PYKCSWEGCE WRFARSDELT RHYRKHTGAK PFKCNHCDRC FSRSDHLALH MKRHI |
| Buffer | The KLF7 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction