missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLF6 Protein
Specifications
Specifications
| Name | KLF6 Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMDVLPMC SIFQELQIVH ETGYFSALPS LEEYWQQTCL ELERYLQSEP CYVSASEIKF DSQEDLWTKI ILAREKKEES ELKISSSPPE DTLISPSFCY NLETNSLNSD VSSESSDSSE ELSPTAKFTS DPIGEVLVSS GKLSSSVTST PPSSPELSRE PSQLWGCVPG ELPSPGKVRS GTSGKPGDKG NGDASPDGRR RVHRCHFNGC RKVYTKSSHL KAHQRTHTGE KPYRCSWEGC EWRFARSDEL TRHFRKHTGA KPFKCSHCDR CFSRSDHLAL HMKRHL |
| Buffer | The KLF6 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?