missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KLF4 Protein

A cDNA sequence encoding the KLF4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12498-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15940609

  • £155.00 / 10µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

KLF4 Protein
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMAVS DALLPSFSTF ASGPAGREKT LRQAGAPNNR WREELSHMKR LPPVLPGRPY DLAAATVATD LESGGAGAAC GGSNLAPLPR RETEEFNDLL DLDFILSNSL THPPESVAAT VSSSASASSS SSPSSSGPAS APSTCSFTYP IRAGNDPGVA PGGTGGGLLY GRESAPPPTA PFNLADINDV SPSGGFVAEL LRPELDPVYI PPQQPQPPGG GLMGKFVLKA SLSAPGSEYG SPSVISVSKG SPDGSHPVVV APYNGGPPRT CPKIKQEAVS SCTHLGAGPP LSNGHRPAAH DFPLGRQLPS RTTPTLGLEE VLSSRDCHPA LPLPPGFHPH PGPNYPSFLP DQMQPQVPPL HYQELMPPGS CMPEEPKPKR GRRSWPRKRT AT
Greater than 85% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
KLF4 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers