missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KISS1 Protein
A cDNA sequence encoding the KISS1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12494-1mg
Additional Details : Weight : 0.01000kg
Specifications
KISS1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MEPLEKVASV GNSRPTGQQL ESLGLLAPGE QSLPCTERKP AATARLSRRG TSLSPPPESS GSPQQPGLSA PHSRQIPAPQ GAVLVQREKD LPNYNWNSFG LRFGKREAAP GNHGRSAGRG | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein solution (1 mg/ml) contains 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol. |