missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human KIR3DL3/CD158z Fc Chimera Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human B7-H7/HHLA2 His-tag Protein, CF (Catalog # 10475-B7 ) is immobilized at 2 μg/mL (100 μL/well), the concentration of Recombinant Human KIR3DL3/CD158z Fc Chimera (Catalog # 10104-KR) that produces 50% of the optimal binding response is found to be approximately 45.0-270 ng/mL.
Specifications
Specifications
| Accession Number | NP_703144.2 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose |
| Gene ID (Entrez) | 115653 |
| Molecular Weight (g/mol) | M.W.-observed: 62-71 kDa, under reducing conditions, M.W.-theroretical: 59 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human KIR3DL3/CD158z protein Human KIR3DL3/CD158z (Gln26-Leu322) (N-terminus) Accession # NP_703144.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction