missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KIR2DL3 Protein
A cDNA sequence encoding the KIR2DL3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12488-20ug
Additional Details : Weight : 0.01000kg
Specifications
KIR2DL3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MEGVHRKPSL LAHPGPLVKS EETVILQCWS DVRFQHFLLH REGKFKDTLH LIGEHHDGIS KANFSIGPMM QDLAGTYRCY GSVTHSPYQL SAPSDPLDIV ITGLYEKPSL SAQPGPTVLA GESVTLSCSS RSSYDMYHLS REGEAHERRF SAGPKVNGTF QADFPLGPAT HGGTYRCFGS FRDSPYEWSN SSDPLLVSVT GN | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein (1 mg/ml) contains 25mM Tris-HCl (pH 7.5). |