missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KIR2DL3 Protein

A cDNA sequence encoding the KIR2DL3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12488-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15940579

  • £3965.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

KIR2DL3 Protein
Research Use Only
Recombinant Protein
E. coli
MEGVHRKPSL LAHPGPLVKS EETVILQCWS DVRFQHFLLH REGKFKDTLH LIGEHHDGIS KANFSIGPMM QDLAGTYRCY GSVTHSPYQL SAPSDPLDIV ITGLYEKPSL SAQPGPTVLA GESVTLSCSS RSSYDMYHLS REGEAHERRF SAGPKVNGTF QADFPLGPAT HGGTYRCFGS FRDSPYEWSN SSDPLLVSVT GN
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The protein (1 mg/ml) contains 25mM Tris-HCl (pH 7.5).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers