missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KHDC1L Protein
A cDNA sequence encoding the KHDC1L was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12484-5ug
Additional Details : Weight : 0.01000kg
Specifications
KHDC1L Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAVGTSA LSKEPWWTLP ENFHSPMVFH MEEDQEELIF GLDDTYLRCI ELHSHTLIQL ERCFTATGQT RVTVVGPPMA KQWLLLMFHC VGSQDSKCHA RGLKMLERVR SQPLTNDDLV TSVSLPPYTG D | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
KHDC1L protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT. |