missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human KGF/FGF-7 Protein
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA.
Unit Size:
10µg
50µg
Description
Measured in a cell proliferation assay using 4MBr-5 rhesus monkey epithelial cells. Rubin, J.S. et al. (1989) Proc. Natl. Acad. Sci. USA 86:802. The ED50 for this effect is typically 6-60 ng/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA with BSA as a carrier protein. |
| Gene ID (Entrez) | 2252 |
| Molecular Weight (g/mol) | 19 kDa |
| Name | KGF/FGF-7 |
| Quantity | 10 μg |
| Source | E. coli-derived human KGF/FGF-7 protein Cys32-Thr194,with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | FGF7, FGF-7, fibroblast growth factor 7HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor) |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction