missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Keratin, type II cytoskeletal 4 Protein

A DNA sequence encoding the Homo sapiens (Human) Keratin, type II cytoskeletal 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6281-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16072160

  • £1265.00 / 500µg
Estimated Shipment: 15-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Keratin, type II cytoskeletal 4 Protein
Research Use Only
KRT4
Recombinant Protein
Yeast
MIARQQCVRG GPRGFSCGSA IVGGGKRGAF SSVSMSGGAG RCSSGGFGSR SLYNLRGNKS ISMSVAGSRQ GACFGGAGGF GTGGFGAGGF GAGFGTGGFG GGFGGSFSGK GGPGFPVCPA GGIQEVTINQ SLLTPLHVEI DPEIQKVRTE EREQIKLLNN KFASFIDKVQ FLEQQNKVLE TKWNLLQQQT TTTSSKNLEP LFETYLSVLR KQLDTLGNDK GRLQSELKTM QDSVEDFKTK YEEEINKRTA AENDFVVLKK DVDAAYLNKV ELEAKVDSLN DEINFLKVLY DAELSQMQTH VSDTSVVLSM DNNRNLDLDS IIAEVRAQYE EIAQRSKAEA EALYQTKVQQ LQISVDQHGD NLKNTKSEIA ELNRMIQRLR AEIENIKKQC QTLQVSVADA EQRGENALKD AHSKRVELEA ALQQAKEELA RMLREYQELM SVKLALDIEI ATYRKLLEGE EYRMSGECQS AVSISVVSGS TSTGGISGGL GSGSGFGLSS GFGSGSGSGF GFGGSVSGSS SSKIISTTTL NKRR
Greater than 90% as determined by SDS-PAGE.
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers