missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Kallikrein 1 Protein

Product Code. 18106640
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18106640

Brand: R&D Systems™ 2337SE010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Human Kallikrein 1 Protein is derived from NS0. The Recombinant Human Kallikrein 1 Protein has been validated for the following applications: Enzyme Activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and CaCl 2 .
Gene ID (Entrez) 3816
Molecular Weight (g/mol) 32 kDa
Name Kallikrein 1
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived human Kallikrein 1 protein Ile25-Ser262,with an N-terminal human KLK-5 propeptide (Val23-Arg66) and a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Structural Form Pro form
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.