missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human JNK2 / MAPK9 Protein

A DNA sequence encoding the Homo sapiens (Human) JNK2 / MAPK9, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6342-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022610

  • £1735.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001128516.1
JNK2 / MAPK9 Protein
Research Use Only
MAPK9
Recombinant Protein
E. coli
MSDSKCDSQF YSVQVADSTF TVLKRYQQLK PIGSGAQGIV CAAFDTVLGI NVAVKKLSRP FQNQTHAKRA YRELVLLKCV NHKNIISLLN VFTPQKTLEE FQDVYLVMEL MDANLCQVIH MELDHERMSY LLYQMLCGIK HLHSAGIIHR DLKPSNIVVK SDCTLKILDF GLARTACTNF MMTPYVVTRY YRAPEVILGM GYKENVDIWS VGCIMGELVK GCVIFQGTDH IDQWNKVIEQ LGTPSAEFMK KLQPTVRNYV ENRPKYPGIK FEELFPDWIF PSESERDKIK TSQARDLLSK MLVIDPDKRI SVDEALRHPY ITVWYDPAEA EAPPPQIYDA QLEEREHAIE EWKELIYKEV MDWEERSKNG VVKDQPSDAA VSSNATPSQS SSINDISSMS TEQTLASDTD SSLDASTGPL EGCR
Greater than 90% as determined by SDS-PAGE.
5601
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers