missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IRF 2 Protein

A cDNA sequence encoding the IRF 2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12457-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15940479

  • £155.00 / 20µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

3660
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MPVERMRMRP WLEEQINSNT IPGLKWLNKE KKIFQIPWMHAARHGWDVEK DAPLFRNWAI HTGKHQPGVD KPDPKTWKAN FRCAMNSLPD IEEVKDKSIKKGNNAFRVYR MLP
Greater than 95.0% as determined by SDS-PAGE.
IRF 2 Protein
Research Use Only
IRF2
Human
His
1 mg/ml in 20mM Tris pH 8.0, 1mM DTT and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers