missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IP6K2 Protein
A cDNA sequence encoding the IP6K2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12454-2ug
Additional Details : Weight : 0.01000kg
Specifications
IP6K2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSPAFRAMDV EPRAKGVLLE PFVHQVGGHS CVLRFNETTL CKPLVPREHQ FYETLPAEMR KFTPQYKGVV SVRFEEDEDR NLCLIAYPLK GDHGIVDIVD NSDCEPKSKL LRWTTNKKHH VLETEKTPKD WVRQHRKEEK MKSHKLEEEF EWLKKSEVLY YTVEKKGNIS SQLKHYNPWS MKCHQQQLQR MKENAKHRNQ YKFILLENLT SRYEVPCVLD LKMGTRQHGD DASEEKAANQ IRKCQQSTSA VIGVRVCGMQ VYQAGSGQLM FMNKYHGRKL SVQGFKEALF QFFHNGRYLR RELLGPVLKK LTELKAVLER QESYRFYSSS LLVIYDGKER PEVVLDSDAE DLEDLSEESA DESAGAYAYK PIGASSVDVR MIDFAHTTCR LYGEDTVVHE GQDAGYIFGL QSLIDIVTEI SEESGE | |
Greater than 85% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
IP6K2 solution (0.5 mg/ml) contains Tris-HCl buffer (pH 8.0) and 10% glycerol. |