missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Interferon-induced transmembrane protein 1

A DNA sequence encoding the Homo sapiens (Human) Interferon-induced transmembrane protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8354-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16074181

  • £384.00 / 100µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003632.3
Interferon-induced transmembrane protein 1
Research Use Only
IFITM1
Recombinant Protein
E. coli
MHKEEHEVAV LGPPPSTILP RSTVINIHSE TSVPDH
Greater than 90% as determined by SDS-PAGE.
8519
100 μg
Not determined.
Extracellular Domain
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers