missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interferon-induced transmembrane protein 1
A DNA sequence encoding the Homo sapiens (Human) Interferon-induced transmembrane protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8354-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003632.3 | |
Interferon-induced transmembrane protein 1 | |
Research Use Only | |
IFITM1 | |
Recombinant Protein | |
E. coli | |
MHKEEHEVAV LGPPPSTILP RSTVINIHSE TSVPDH | |
Greater than 90% as determined by SDS-PAGE. |
8519 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |