missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Integrin alpha X beta 2 Protein
Description
The Recombinant Human Integrin alpha X beta 2 Protein is derived from CHO. The Recombinant Human Integrin alpha X beta 2 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 3687 |
| Molecular Weight (g/mol) | 128.9 kDa ( alphaX) & 83.1 kDa (beta2) |
| Name | Integrin alpha X beta 2 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha X beta 2 protein, (N-terminus) Human Integrin alpha X (Phe20-Pro1107) Accession &Num; P20702 Acidic Tail 6-His tag Human Integrin beta 2 (Gln23-Asn700) Accession &Num; AAA59490 Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?