missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Integrin alpha V beta 5 Protein

Product Code. 18359667
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359667 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359667 Supplier R&D Systems™ Supplier No. 2528AV050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Integrin alpha V beta 5 Protein is derived from CHO. The Recombinant Human Integrin alpha V beta 5 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 3685
Molecular Weight (g/mol) 115.4 kDa ( alphaV subunit) & 84.8 kDa (beta5 subunit)
Name Integrin alpha V beta 5
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha V beta 5 protein, (N-terminus) Human Integrin alpha V (Phe31-Val992) Accession &Num; P06756 His-Pro GGGSGGGS Acidic Tail 6-His tag Human Integrin beta 5 (Gly24-Asn719) Accession &Num; P18084 His-Pro GGGSGGGS Basic Tail (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Noncovalently-linked heterodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.