missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Integrin alpha V beta 3 Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Human Integrin alpha V beta 3 Protein is derived from CHO. The Recombinant Human Integrin alpha V beta 3 Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and CaCl 2 . |
| Gene ID (Entrez) | 3685 |
| Molecular Weight (g/mol) | 110.5 kDa ( alphaV subunit), 80.8 kDa (beta3 subunit) |
| Name | Integrin alpha V beta 3 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha V beta 3 protein, (N-terminus) Human Integrin alpha V subunit (Phe31-Val992) Accession &Num; NP_002201 His-Pro GGGSGGGS Acidic Tail Human Integrin beta 3 subunit (Gly27-Asp718) Accession &Num; AAA52589 His-Pro GGGSGGGS Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction