missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Integrin alpha 3 beta 1/VLA-3 Protein
Description
The Recombinant Human Integrin alpha 3 beta 1/VLA-3 Protein is derived from CHO. The Recombinant Human Integrin alpha 3 beta 1/VLA-3 Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl. |
| Gene ID (Entrez) | 3675 |
| Molecular Weight (g/mol) | Observed Molecular Wt: 150kDa and 125kDa, reducing conditions, Predicted Molecular Wt.: 110.5kDa (Integrin alpha 3), 82.3kDa (Integrin beta 1) |
| Name | Integrin alpha 3 beta 1/VLA-3 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha 3 beta 1/VLA-3 protein, (N-terminus) Human Integrin alpha 3 (Phe33-Glu991) Accession &Num; P26006 acidic tail Human Integrin beta 1 (Glu21-Asp728) Accession &Num; P05556 basic tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?