missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Integrin alpha 10 beta 1 Protein

Product Code. 18177770
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18177770

Brand: R&D Systems™ 5895AB050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Mouse Laminin I (Catalog # 3400-010-02 ) is coated at 10 μg/mL, Recombinant Human Integrin alpha 10 beta 1 binds with an apparent Kd <5 nM.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75578 (Integrin alpha 10) & P05556 (Integrin beta 1)
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 8515
Molecular Weight (g/mol) M.W. (Observed): 160-175 kDa & 125-150 kDa, reducing conditions; M.W. (theoretical): 128.8 kDa (alpha 10) & 86.4 kDa (beta 1)
Name Integrin alpha 10 beta 1
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Integrin alpha 10 beta 1 protein Human Integrin alpha 10 (Phe23-Ser1122) (N-terminus) Accession # O75578GGGSGGGSGGGS Acidic Tail Human Integrin ss1 (Gln21-Asp728) (N-terminus) Accession # P05556 GGGSGGGSGGGS Basic Tail (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Integrin alpha 10 beta 1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.