missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Integrin alpha 1 beta 1 Protein

Product Code. 18118380
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18118380

Brand: R&D Systems™ 7064AB025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Integrin alpha 1 beta 1 Protein is derived from CHO. The Recombinant Human Integrin alpha 1 beta 1 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 3672
Molecular Weight (g/mol) 132.3 kDa (Integrin alpha1) & 86.4 kDa (Integrin beta1)
Name Integrin alpha 1 beta 1
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha 1 beta 1 protein, (N-terminus) Human Integrin alpha 1 (Met1-Pro1141) Accession &Num; P56199 GGGSGGGSGGGS Acidic Tail HHHHHH Human Integrin beta 1 (Gln21-Asp728) Accession &Num; P05556 GGGSGGGSGGGS Basic Tail (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.