missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Insulin R/CD220 (aa 28-956) Protein

Product Code. 18708318
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18708318

Brand: R&D Systems™ 8974IR050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Insulin R/CD220 (aa 28-956) Protein is derived from HEK293. The Recombinant Human Insulin R/CD220 (aa 28-956) Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 3643
Molecular Weight (g/mol) Observed M.W.: 105-120kDa ( alpha subunit) and 37-49kDa ( beta subunit), reducing conditions, Theoretical M.W.: 107kDa ( alpha & beta heterodimer)
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human Insulin R/CD220 protein His28-Arg762( alpha subunit) and Ser763-Lys956 with a c-terminal 10-His tag ( beta subunit)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, INSR, insulin receptor, InsulinR, IR
Common Name Insulin R/CD220
Gene Symbol INSR
Biological Activity Measured by its binding ability in a functional ELISA. When 15ng/mL of biotinylated recombinant human Insulin is added to serially diluted Recombinant Human Insulin R/CD220 B Isoform, the concentration of Recombinant Human Insulin R/CD220 B Isoform that produces 50% of the optimal binding response is typically 0.1-0.5μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Human Insulin R alpha (His28-Arg762) Accession # P06213, N-terminus; Human Insulin R beta (Ser763-Lys956) Accession # P06213; HHHHHHHHHH, C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.