missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Insulin R/CD220 (aa 28-944) Protein

Product Code. 18188300
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18188300

Brand: R&D Systems™ 1544IR050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Human Insulin R/CD220 (aa 28-944) Protein is derived from NS0. The Recombinant Human Insulin R/CD220 (aa 28-944) Protein has been validated for the following applications: Binding Activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 3643
Molecular Weight (g/mol) 82.9 kDa ( alpha subunit), 22.9 kDa (beta subunit)
Name Insulin R/CD220
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human Insulin R/CD220 protein His28-Arg750 ( alpha subunit) & Ser751-Lys944 with a C-terminal 10-His tag ( beta subunit)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Tetramer; disulfide-linked homodimer of disulfide-linked heterodimers ( alpha & beta )
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.