missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human ING2 Protein

Product Code. 18236853 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
100 μg
Unit Size:
0.50mg
100µg
This item is not returnable. View return policy

Product Code. 18236853

Brand: Novus Biologicals™ NBP261168100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids1-280 of Human ING2 The Recombinant Human ING2 Protein is derived from E. coli. The Recombinant Human ING2 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/ml
For Use With (Application) SDS-PAGE
Formulation 20mM MES (pH5.0) containing 20% Glycerol, 0.1M NaCl
Gene ID (Entrez) 3622
Molecular Weight (g/mol) 35.2 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol ING2
Cross Reactivity ING2
Species Human
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.