missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Indian Hedgehog/Ihh C28II Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
Indian Hedgehog Protein is fully biologically active when compared to standard. The ED50 as determined by its ability to induce alkaline phosphatase production by C3H10T1/2(CCL-226) cells is 3.0-10 μg/mL.
Specifications
Specifications
| Accession Number | Q14623 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 1 x PBS, 0.02% Tween-20, pH 7.0. |
| Gene ID (Entrez) | 3549 |
| Molecular Weight (g/mol) | 19.8 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of Indian Hedgehog/Ihh as determined by LAL method. |
| Gene Alias | BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction