missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IMP3 Protein

A cDNA sequence encoding the IMP3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12431-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15940399

  • £2975.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

IMP3 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMVRKLKF HEQKLLKQVD FLNWEVTDHN LHELRVLRRY RLQRREDYTR YNQLSRAVRE LARRLRDLPE RDQFRVRASA ALLDKLYALG LVPTRGSLEL CDFVTASSFC RRRLPTVLLK LRMAQHLQAA VAFVEQGHVR VGPDVVTDPA FLVTRSMEDF VTWVDSSKIK RHVLEYNEER DDFDLEA
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
IMP3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers