missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Immortalization up-regulated Protein
A DNA sequence encoding the Homo sapiens (Human) Immortalization up-regulated protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5733-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001304730.1 | |
Immortalization up-regulated Protein | |
Research Use Only | |
IMUP | |
Recombinant Protein | |
E. coli | |
MEFDLGAALE PTSQKPGVGA GHGGDPKLSP HKVQGRSEAG AGPGPKASNF RGLGKGRRLT AAPPSSKDTT ALPTPAAAPA IRTRM | |
Greater than 90% as determined by SDS-PAGE. |
64073 | |
1 mg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |