missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL34 Protein
A cDNA sequence encoding the IL34 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10728-10ug
Additional Details : Weight : 0.01000kg
Specifications
146433 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The specific activity was measured by its ability to induce secretion of MCP-1 from PBMCs. | |
Human | |
His | |
The IL34 was lyophilized from a solution containing 20mM Sodium Citrate, pH 3.0 and 75mM NaCl. |
IL34 Protein | |
Research Use Only | |
IL34 | |
Recombinant Protein | |
Human (HEK293) | |
NEPLEMWPLT QNEECTVTGF LRDKLQYRSR LQYMKHYFPI NYKISVPYEG VFRIANVTRL QRAQVSEREL RYLWVLVSLS ATESVQDVLL EGHPSWKYLQ EVETLLLNVQ QGLTDVEVSP QGLTDVEVSP APGPNLKLVR PKALLDNCFR VMELLYCSCC KQSSVLNWQD CEVPSPQSCS PEPSLQYAAT QLYPPPPWSP SSPPHSTGSV RPVRAQGEGL LPHHHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |