missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL28B Protein
A cDNA sequence encoding the IL28B was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Gene ID (Entrez) | 282617 |
---|---|
Name | IL28B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | IFNL3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15940359
|
enQuireBio™
QP12417-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920359
|
enQuireBio™
QP12417-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930359
|
enQuireBio™
QP12417-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
282617 | |
Research Use Only | |
IFNL3 | |
Human | |
His | |
IL 28B protein solution (0.25 mg/ml) contains Phosphate buffered saline (pH 7.4), 20% glycerol, 1mM EDTA and 0.1mM PMSF. |
IL28B Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
VPVARLRGAL PDARGCHIAQ FKSLSPQELQ AFKRAKDALE ESLLLKDCKC RSRLFPRTWD LRQLQVRERP VALEAELALT LKVLEATADT DPALGDVLDQ PLHTLHHILS QLRACIQPQP TAGPRTRGRL HHWLHRLQEA PKKESPGCLE ASVTFNLFRL LTRDLNCVAS GDLCVHHHHHH | |
Greater than 85.0% as determined by SDS-PAGE. |