missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL25 Protein

Recombinant Protein

Brand:  enQuireBio™ QP5273-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955819

  • £1890.00 / 500µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_073626.1
IL25 Protein
Research Use Only
IL25
Recombinant Protein
E. coli
MYSHWPSCCP SKGQDTSEEL LRWSTVPVPP LEPARPNRHP ESCRASEDGP LNSRAISPWR YELDRDLNRL PQDLYHARCL CPHCVSLQTG SHMDPRGNSE LLYHNQTVFY RRPCHGEKGT HKGYCLERRL YRVSLACVCV RPRVMG
Greater than 95% as determined by SDS-PAGE.
64806
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Human IL25 induces IL-8 production in human PBMCs.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers