missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL25 Protein
Recombinant Protein
Marque: enQuireBio™ QP5273-100ug
Informations supplémentaires : Weight : 0.01000kg
Spécification
NP_073626.1 | |
IL25 Protein | |
Research Use Only | |
IL25 | |
Recombinant Protein | |
E. coli | |
MYSHWPSCCP SKGQDTSEEL LRWSTVPVPP LEPARPNRHP ESCRASEDGP LNSRAISPWR YELDRDLNRL PQDLYHARCL CPHCVSLQTG SHMDPRGNSE LLYHNQTVFY RRPCHGEKGT HKGYCLERRL YRVSLACVCV RPRVMG | |
Greater than 95% as determined by SDS-PAGE. |
64806 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human IL25 induces IL-8 production in human PBMCs. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |