missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL1RAPL2 Fc Chimera Protein

Product Code. 18399667
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399667 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399667 Supplier R&D Systems™ Supplier No. 1007MR100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Human IL1RAPL2 Fc Chimera is immobilized at 1μg/well, the concentration of Recombinant Human IL-1 Rrp2/IL-1 R6 Fc Chimera (Catalog # 872-RP ) that produces 50% of the optimal binding response is found to be approximately 2-8μg/mL.

The Recombinant Human IL-1 RAPL2/IL-1 R9 Fc Chimera Protein is derived from NS0. The Recombinant Human IL-1 RAPL2/IL-1 R9 Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 26280
Molecular Weight (g/mol) 66 kDa (monomer)
Name NPRA/NPR1
Quantity 100 μg
Source Mouse myeloma cell line,NS0-derived human IL1RAPL2 protein, (N-terminus) Human IL1RAPL2 (Thr17-Glu356) & (Lys20-Glu356) Accession &Num; Q9NP60 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Gene Alias IL-1R10, IL-1R-9, IL1R9IL-1 receptor accessory protein-like 2, IL-1R9IL-1RAPL-2, IL1RAPL2, IL-1RAPL2, IL-1-RAPL-2, IL1RAPL-2IL1RAPL-2-related protein, interleukin 1 receptor 9, interleukin 1 receptor accessory protein-like 2, Three immunoglobulin domain-containing IL-1 receptor-related 1, TIGIRR-1, TIGIRR-1Interleukin-1 receptor 9, X-linked interleukin-1 receptor accessory protein-like 2
Common Name NPRA/NPR1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.