missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL1R1 Protein
A cDNA sequence encoding the IL1R1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12412-2ug
Additional Details : Weight : 0.01000kg
Specifications
3554 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
DKCKEREEKI ILVSSANEID VRPCPLNPNE HKGTITWYKD DSKTPVSTEQ ASRIHQHKEK LWFVPAKVED SGHYYCVVRN SSYCLRIKIS AKFVENEPNL CYNAQAIFKQ KLPVAGDGGL VCPYMEFFKN ENNELPKLQW YKDCKPLLLD NIHFSGVKDR LIVMNVAEKH RGNYTCHASY TYLGKQYPIT RVIEFITLEE NKPTRPVIVS PANETMEVDL GSQIQLICNV TGQLSDIAYW KWNGSVIDED DPVLGEDYYS VENPANKRRS TLITVLNISE IESRFYKHPF TCFAKNTHGI DAAYIQLIYP VTNFQKLEHH HHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
IL1R1 Protein | |
Research Use Only | |
IL1R1 | |
Human | |
His | |
IL1R1 protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |