missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL1R1 Protein

A cDNA sequence encoding the IL1R1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12412-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960339

  • £155.00 / 10µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

3554
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Baculovirus
DKCKEREEKI ILVSSANEID VRPCPLNPNE HKGTITWYKD DSKTPVSTEQ ASRIHQHKEK LWFVPAKVED SGHYYCVVRN SSYCLRIKIS AKFVENEPNL CYNAQAIFKQ KLPVAGDGGL VCPYMEFFKN ENNELPKLQW YKDCKPLLLD NIHFSGVKDR LIVMNVAEKH RGNYTCHASY TYLGKQYPIT RVIEFITLEE NKPTRPVIVS PANETMEVDL GSQIQLICNV TGQLSDIAYW KWNGSVIDED DPVLGEDYYS VENPANKRRS TLITVLNISE IESRFYKHPF TCFAKNTHGI DAAYIQLIYP VTNFQKLEHH HHHH
Greater than 95.0% as determined by SDS-PAGE.
IL1R1 Protein
Research Use Only
IL1R1
Human
His
IL1R1 protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers