missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL12 Protein
A cDNA sequence encoding the IL12 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | IL12 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The activity is determined by the IFN-g ELISA in a using NK-92 cell. The ED50 for this effect is less than or equal to 0.03 ng/ml. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974968
|
enQuireBio™
QP10722-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954968
|
enQuireBio™
QP10722-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964968
|
enQuireBio™
QP10722-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IL12 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Baculovirus | |
IL12A(p35):RNLPVATPDP GMFPCLHHSQ NLLRAVSNML QKARQTLEFY PCTSEEIDHE DITKDKTSTV EACLPLELTK NESCLNSRET SFITNGSCLA SRKTSFMMAL CLSSIYEDLK MYQVEFKTMN AKLLMDPKRQ IFLDQNMLAV IDELMQALNF NSETVPQKSS LEEPDFYKTK IKLCILLHAF RIRAVTIDRV MSYLNASHHH HHH IL12B(p40):IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS | |
Greater than 90.0% as determined by SDS-PAGE. |
Research Use Only | |
The activity is determined by the IFN-g ELISA in a using NK-92 cell. The ED50 for this effect is less than or equal to 0.03 ng/ml. | |
Human | |
His | |
IL12 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol. |