missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human IL-9 Protein

Product Code. 18497631 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18497631 100 μg 100µg
18418571 10 μg 10µg
18428281 1 mg 1mg
18487631 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18497631 Supplier Novus Biologicals™ Supplier No. NBP234909100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

IL9 Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human MO7e cells is less than 0.2 ng/mL, corresponding to a specific activity of >5.0 x 106IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P15248, P15248, P15248
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
Gene ID (Entrez) 3578
Molecular Weight (g/mol) 14.1 kDa
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of IL-9 as determined by LAL method.
Gene Alias Cytokine P40, HP40, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40
Gene Symbol IL9
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein IL-9
Show More Show Less
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.